ENA423209757 Enamine

4-chloro-1-(piperidin-1-yl)butan-1-one

  • MDL number MFCD00544305
  • Cas Number : 90436-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171220

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171220
Availability details for
Quantity
  • Availability
  • Add to Cart