ENA423209760 Enamine

ethyl(1H-indol-5-ylmethyl)amine

  • MDL number MFCD13875546
  • Cas Number : 946680-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171298

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171298
Availability details for
Quantity
  • Availability
  • Add to Cart