ENA423209761 Enamine

2-(bromomethyl)-1-oxaspiro[4.5]decane

  • MDL number MFCD27980901
  • Cas Number : 117038-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171299

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171299
Availability details for
Quantity
  • Availability
  • Add to Cart