ENA423209772 Enamine

N-(2-aminophenyl)-1,1,1-trifluoromethanesulfonamide

  • MDL number MFCD12086253
  • Cas Number : 53718-45-9
  • Molecular Weight : 240.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171318
Availability details for
Quantity
  • Availability
  • Add to Cart