ENA423209775 Enamine

3-fluoropyrrolidine-3-carbonitrile hydrochloride

  • MDL number MFCD21607329
  • Cas Number : 1363405-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171322
Availability details for
Quantity
  • Availability
  • Add to Cart