ENA423209782 Enamine

1-(chloromethyl)-4-[2-(2-methoxyethoxy)ethoxy]benzene

  • MDL number MFCD19672600
  • Cas Number : 314286-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171344

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171344
Availability details for
Quantity
  • Availability
  • Add to Cart