ENA423209792 Enamine

4-(methylsulfanyl)-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD00127853
  • Cas Number : 6958-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171371

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171371
Availability details for
Quantity
  • Availability
  • Add to Cart