ENA423209800 Enamine

1-(3-amino-3-methylazetidin-1-yl)ethan-1-one

  • MDL number MFCD21325269
  • Cas Number : 1480257-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171408
Availability details for
Quantity
  • Availability
  • Add to Cart