ENA423209803 Enamine

1-(1,3-dioxaindan-5-yl)-2-(benzylamino)ethan-1-ol

  • MDL number MFCD11928801
  • Cas Number : 21784-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171424
Availability details for
Quantity
  • Availability
  • Add to Cart