ENA423209805 Enamine

N-[(3-bromophenyl)methyl]oxan-4-amine

  • MDL number MFCD12088387
  • Cas Number : 1155634-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171427

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171427
Availability details for
Quantity
  • Availability
  • Add to Cart