ENA423209808 Enamine

2-(iodomethyl)benzonitrile

  • MDL number MFCD20415038
  • Cas Number : 871892-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171441
Availability details for
Quantity
  • Availability
  • Add to Cart