ENA423209817 Enamine

N-[(3-chlorophenyl)methyl]-2-cyanoacetamide

  • MDL number MFCD02111799
  • Cas Number : 64488-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171459
Availability details for
Quantity
  • Availability
  • Add to Cart