ENA423209820 Enamine

2-cyano-N-[(3,4-difluorophenyl)methyl]acetamide

  • MDL number MFCD14684098
  • Cas Number : 566926-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171462
Availability details for
Quantity
  • Availability
  • Add to Cart