ENA423209823 Enamine

methyl 2-(methylsulfamoyl)benzoate

  • MDL number MFCD11646542
  • Cas Number : 145889-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171465

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-171465
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart