ENA423209827 Enamine

1-methyl-4-phenyl-1H-pyrazol-3-amine

  • MDL number MFCD20483751
  • Cas Number : 30823-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171495
Availability details for
Quantity
  • Availability
  • Add to Cart