ENA423209837 Enamine

(4-butoxy-3-chlorophenyl)methanamine

  • MDL number MFCD14656261
  • Cas Number : 744971-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-171526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171526
Availability details for
Quantity
  • Availability
  • Add to Cart