ENA423209846 Enamine

2-{1-[(4-methoxyphenyl)methyl]-1H-1,2,3-triazol-4-yl}ethan-1-ol

  • MDL number MFCD19108095
  • Cas Number : 1270874-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171545
Availability details for
Quantity
  • Availability
  • Add to Cart