ENA423209847 Enamine

2-(3-methylphenyl)-2-(morpholin-4-yl)acetonitrile

  • MDL number MFCD10005102
  • Cas Number : 66549-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171559
Availability details for
Quantity
  • Availability
  • Add to Cart