ENA423209860 Enamine

4-{8-oxa-3-azabicyclo[3.2.1]octan-3-yl}aniline

  • MDL number MFCD21761899
  • Cas Number : 1416369-78-2
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-171619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171619
Availability details for
Quantity
  • Availability
  • Add to Cart