ENA423209862 Enamine

tert-butyl oxolane-2-carboxylate

  • MDL number MFCD19600043
  • Cas Number : 1342514-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171621
Availability details for
Quantity
  • Availability
  • Add to Cart