ENA423209863 Enamine

tert-butyl oxolane-3-carboxylate

  • MDL number MFCD19600075
  • Cas Number : 186318-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171622

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171622
Availability details for
Quantity
  • Availability
  • Add to Cart