ENA423209873 Enamine

[1-(2-chlorophenyl)ethyl](2-methylpropyl)amine

  • MDL number MFCD11139688
  • Cas Number : 1019578-92-7
  • Molecular Weight : 211.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-169478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-169478
Availability details for
Quantity
  • Availability
  • Add to Cart