ENA423209874 Enamine

3-bromo-N-pentylbenzamide

  • MDL number MFCD01215156
  • Cas Number : 333345-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171640

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171640
Availability details for
Quantity
  • Availability
  • Add to Cart