ENA423209882 Enamine

2,2-dimethyl-1-[4-(trifluoromethyl)phenyl]propan-1-ol

  • MDL number MFCD16316341
  • Cas Number : 109769-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171664

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171664
Availability details for
Quantity
  • Availability
  • Add to Cart