ENA423209884 Enamine

3-ethyl-3-hydroxypentanenitrile

  • MDL number MFCD21801134
  • Cas Number : 17190-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171673
Availability details for
Quantity
  • Availability
  • Add to Cart