ENA423209886 Enamine

1-(3-chloro-2-fluorophenyl)propan-1-ol

  • MDL number MFCD20400977
  • Cas Number : 1343664-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171701

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171701
Availability details for
Quantity
  • Availability
  • Add to Cart