ENA423209894 Enamine

2-cyano-N-[1-(4-methoxyphenyl)ethyl]acetamide

  • MDL number MFCD07166643
  • Cas Number : 894644-29-2
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-cyano-N-[1-(4-methoxyphenyl)ethyl]acetamide
Inchi Code 1S/C12H14N2O2/c1-9(14-12(15)7-8-13)10-3-5-11(16-2)6-4-10/h3-6,9H,7H2,1-2H3,(H,14,15)
Inchi Key ULLRLUHVSBEXDY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-171743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171743
Availability details for
Quantity
  • Availability
  • Add to Cart