ENA423209896 Enamine

2-cyano-N-[(2-methylphenyl)methyl]acetamide

  • MDL number MFCD16044208
  • Cas Number : 566926-04-3
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-171745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171745
Availability details for
Quantity
  • Availability
  • Add to Cart