ENA423209898 Enamine

2-(2-methanesulfinylphenyl)acetic acid

  • MDL number MFCD20641340
  • Cas Number : 90536-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171749
Availability details for
Quantity
  • Availability
  • Add to Cart