ENA423209899 Enamine

oxolan-3-amine hydrochloride

  • MDL number MFCD06738925
  • Cas Number : 204512-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171752

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-171752
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart