ENA423209904 Enamine

1-(4-fluoro-2-nitrophenyl)piperidin-3-ol

  • MDL number MFCD11039141
  • Cas Number : 1386140-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171763

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171763
Availability details for
Quantity
  • Availability
  • Add to Cart