ENA423209907 Enamine

5-(chloromethyl)-2,3-dihydro-1,3,4-thiadiazol-2-one

  • MDL number MFCD19233214
  • Cas Number : 84352-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-88
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171766
Availability details for
Quantity
  • Availability
  • Add to Cart