ENA423209910 Enamine

[3-(dimethylamino)propyl](ethyl)amine

  • MDL number MFCD00051552
  • Cas Number : 19475-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171801

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-171801
pictograms GHS02,GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart