ENA423209912 Enamine

5-(4-chlorobenzoyl)pyrimidine

  • MDL number MFCD13152954
  • Cas Number : 103686-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-171803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171803
Availability details for
Quantity
  • Availability
  • Add to Cart