ENA423209916 Enamine

3-cyclohexyl-2-oxopropanoic acid

  • MDL number MFCD16706684
  • Cas Number : 5962-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171807
Availability details for
Quantity
  • Availability
  • Add to Cart