ENA423209917 Enamine

N-[(2-bromophenyl)methyl]-2-cyanoacetamide

  • MDL number MFCD14661192
  • Cas Number : 566926-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171808

Safety Information

msds https://www.enaminestore.com/catalog/EN300-171808
Availability details for
Quantity
  • Availability
  • Add to Cart