ENA423210011 Enamine

3-[3-(dimethylamino)phenyl]propanoic acid hydrochloride

  • MDL number MFCD27980946
  • Cas Number : 99528-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172482

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172482
Availability details for
Quantity
  • Availability
  • Add to Cart