ENA423210013 Enamine

ethyl 2-(cyclopropylmethyl)-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD27980948
  • Cas Number : 1803599-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172565

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172565
Availability details for
Quantity
  • Availability
  • Add to Cart