ENA423210017 Enamine

N4',N4'-dimethyl-[1,1'-biphenyl]-3,4'-diamine hydrochloride

  • MDL number MFCD27980952
  • Cas Number : 1803591-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172738
Availability details for
Quantity
  • Availability
  • Add to Cart