ENA423210025 Enamine

methyl 2-amino-2-(1-methyl-1H-pyrazol-3-yl)acetate hydrochloride

  • MDL number MFCD27980960
  • Cas Number : 1796890-20-4
  • Molecular Weight : 205.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-amino-2-(1-methyl-1H-pyrazol-3-yl)acetate hydrochloride
Inchi Code 1S/C7H11N3O2.ClH/c1-10-4-3-5(9-10)6(8)7(11)12-2;/h3-4,6H,8H2,1-2H3;1H
Inchi Key WJMISEPYYOBQOD-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-173082

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart