ENA423210027 Enamine

4-(aminomethyl)-2,6-dimethylphenol hydrochloride

  • MDL number MFCD27980962
  • Cas Number : 937-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173112

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173112
Availability details for
Quantity
  • Availability
  • Add to Cart