ENA423210029 Enamine

3-methoxy-1-(piperazin-1-yl)propan-1-one hydrochloride

  • MDL number MFCD27980964
  • Cas Number : 1797202-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173127

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173127
Availability details for
Quantity
  • Availability
  • Add to Cart