ENA423210030 Enamine

1-(4H-1,2,4-triazol-3-yl)cyclobutan-1-amine hydrochloride

  • MDL number MFCD27980965
  • Cas Number : 1797202-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173133
Availability details for
Quantity
  • Availability
  • Add to Cart