ENA423210038 Enamine

2,2-difluoro-1-(3-fluoro-4-methoxyphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD27980973
  • Cas Number : 1803604-95-6
  • Molecular Weight : 241.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2-difluoro-1-(3-fluoro-4-methoxyphenyl)ethan-1-amine hydrochloride
Inchi Code 1S/C9H10F3NO.ClH/c1-14-7-3-2-5(4-6(7)10)8(13)9(11)12;/h2-4,8-9H,13H2,1H3;1H
Inchi Key FVLDYUCDPBXRNC-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-173251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173251
Availability details for
Quantity
  • Availability
  • Add to Cart