ENA423210040 Enamine

1-(1H-imidazol-2-yl)-2-methylpropan-1-amine dihydrochloride

  • MDL number MFCD27980975
  • Cas Number : 1797398-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173291

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173291
Availability details for
Quantity
  • Availability
  • Add to Cart