ENA423210043 Enamine

3-[(1,3-dioxo-2,3,3a,4,7,7a-hexahydro-1H-isoindol-2-yl)amino]benzoic acid

  • MDL number MFCD27980978
  • Cas Number : 1803593-41-0
  • Molecular Weight : 286.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-173413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173413
Availability details for
Quantity
  • Availability
  • Add to Cart