ENA423210055 Enamine

5-methylpyrrolidin-3-ol hydrochloride

  • MDL number MFCD27980990
  • Cas Number : 1803584-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173562
Availability details for
Quantity
  • Availability
  • Add to Cart