ENA423210068 Enamine

2-{[(5-chloro-2-hydroxyphenyl)methyl]amino}benzoic acid

  • MDL number MFCD12462653
  • Cas Number : 857602-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-172220
Availability details for
Quantity
  • Availability
  • Add to Cart