ENA423210070 Enamine

2-cyano-N-[1-(2,4-dichlorophenyl)ethyl]acetamide

  • MDL number MFCD11128724
  • Cas Number : 128173-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172230
Availability details for
Quantity
  • Availability
  • Add to Cart