ENA423210071 Enamine

1-chloroethyl 2,2-dimethylpropanoate

  • MDL number MFCD20483880
  • Cas Number : 40258-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172231
Availability details for
Quantity
  • Availability
  • Add to Cart